Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132409.3 | NP_572637.2 | 3 - 4 | 3421 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |