Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298247.1 | NP_001285176.1 | 2 - 3 | 509 | 57 | |
NM_001298248.1 | NP_001285177.1 | 2 - 3 | 733 | 57 | ||
NM_001298249.1 | NP_001285178.1 | 2 - 3 | 509 | 57 | ||
NM_001298250.1 | NP_001285179.1 | 2 - 3 | 733 | 57 | ||
NM_001298251.1 | NP_001285180.1 | 2 - 3 | 509 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |