Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132723.3 | NP_572951.1 | 6 - 7 | 2114 | 73 | |
GenBank mRNA | AY069093.1 | - | 6 - 7 | 2093 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |