Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3-GEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103513.3 | NP_001096983.2 | 12 - 13 | 4541 | 75 | |
NM_001258746.2 | NP_001245675.1 | 13 - 14 | 4559 | 75 | ||
NM_001347810.1 | NP_001334680.1 | 14 - 15 | 4601 | 75 | ||
NM_001347811.1 | NP_001334681.1 | 14 - 15 | 4601 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |