Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3-GEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103513.3 | NP_001096983.2 | 4 - 5 | 1372 | 97 | |
NM_001258746.2 | NP_001245675.1 | 4 - 5 | 1339 | 97 | ||
NM_001258747.1 | NP_001245676.1 | 4 - 5 | 1348 | 97 | ||
NM_001272630.1 | NP_001259559.1 | 4 - 5 | 1348 | 97 | ||
NM_001272631.1 | NP_001259560.1 | 4 - 5 | 1348 | 97 | ||
NM_001316635.1 | NP_001303564.1 | 4 - 5 | 1348 | 97 | ||
NM_001347809.1 | NP_001334679.1 | 4 - 5 | 1348 | 97 | ||
NM_001347810.1 | NP_001334680.1 | 4 - 5 | 1348 | 97 | ||
NM_001347811.1 | NP_001334681.1 | 4 - 5 | 1348 | 97 | ||
NM_132773.3 | NP_573001.2 | 4 - 5 | 1348 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |