Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ack | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139602.3 | NP_647859.1 | 2 - 3 | 1937 | 63 | |
GenBank mRNA | AF181642.1 | - | 2 - 3 | 1918 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |