Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jon65Aii | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139757.2 | NP_648014.1 | 1 - 2 | 348 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |