Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259745.2 | NP_001246674.1 | 12 - 13 | 6831 | 91 | |
NM_001274657.1 | NP_001261586.1 | 11 - 12 | 7184 | 91 | ||
NM_001274658.1 | NP_001261587.1 | 12 - 13 | 6768 | 91 | ||
NM_001274659.1 | NP_001261588.1 | 12 - 13 | 6953 | 91 | ||
NM_001274660.1 | NP_001261589.1 | 13 - 14 | 6969 | 91 | ||
NM_139981.4 | NP_648238.1 | 11 - 12 | 6656 | 91 | ||
GenBank mRNA | AF299248.1 | - | 10 - 11 | 6603 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |