Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273531.1 | NP_001260460.1 | 4 - 5 | 3598 | 63 | |
NM_001273532.1 | NP_001260461.1 | 4 - 5 | 3619 | 63 | ||
NM_057250.4 | NP_476598.1 | 4 - 5 | 3610 | 63 | ||
NM_057251.4 | NP_476599.1 | 4 - 5 | 3631 | 63 | ||
GenBank mRNA | AF145679.1 | - | 4 - 5 | 3532 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |