Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shaw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144310.1 | NP_001137782.1 | 4 - 5 | 785 | 59 | |
NM_001273071.1 | NP_001260000.1 | 5 - 6 | 1043 | 59 | ||
NM_001273072.1 | NP_001260001.1 | 7 - 8 | 1197 | 59 | ||
NM_057373.3 | NP_476721.1 | 7 - 8 | 1197 | 59 | ||
NM_164554.3 | NP_722937.2 | 7 - 8 | 1197 | 59 | ||
GenBank mRNA | AY071475.1 | - | 7 - 8 | 1195 | 59 | |
M32661.1 | - | 4 - 5 | 740 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |