Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057378.3 | NP_476726.2 | 3 - 4 | 2915 | 78 | |
GenBank mRNA | BT023880.1 | - | 3 - 4 | 2911 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |