Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: w | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057439.2 | NP_476787.1 | 2 - 3 | 509 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |