Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pigeon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201915.2 | NP_001188844.1 | 3 - 4 | 857 | 77 | |
NM_057598.5 | NP_476946.1 | 3 - 4 | 857 | 77 | ||
GenBank mRNA | AY061465.1 | - | 3 - 4 | 846 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |