Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299624.1 | NP_001286553.1 | 12 - 13 | 1804 | 60 | |
NM_057661.3 | NP_477009.1 | 12 - 13 | 1807 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |