Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057661.3 | NP_477009.1 | 6 - 7 | 655 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |