Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273836.1 | NP_001260765.1 | 3 - 4 | 1920 | 69 | |
NM_057744.3 | NP_477092.1 | 3 - 4 | 1920 | 69 | ||
GenBank mRNA | BT044167.1 | - | 3 - 4 | 1899 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |