Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273638.1 | NP_001260567.1 | 1 - 2 | 571 | 67 | |
NM_057993.4 | NP_477341.2 | 1 - 2 | 571 | 67 | ||
GenBank mRNA | BT057997.1 | - | 1 - 2 | 570 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |