Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acj6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014743.3 | NP_001014743.1 | 2 - 3 | 907 | 91 | |
NM_001258753.2 | NP_001245682.1 | 2 - 3 | 907 | 91 | ||
NM_080137.3 | NP_524876.1 | 2 - 3 | 907 | 91 | ||
GenBank mRNA | AF086816.1 | - | 2 - 3 | 297 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |