Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aPKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043076.3 | NP_001036541.1 | 8 - 9 | 1952 | 71 | |
NM_001043077.3 | NP_001036542.1 | 6 - 7 | 1591 | 71 | ||
NM_001043078.2 | NP_001036543.1 | 6 - 7 | 1629 | 71 | ||
NM_001202010.4 | NP_001188939.1 | 8 - 9 | 3120 | 71 | ||
NM_001202012.2 | NP_001188941.1 | 7 - 8 | 1751 | 71 | ||
NM_001274055.1 | NP_001260984.1 | 8 - 9 | 3141 | 71 | ||
NM_001274056.2 | NP_001260985.1 | 6 - 7 | 1490 | 71 | ||
NM_001274057.2 | NP_001260986.1 | 6 - 7 | 1591 | 71 | ||
NM_001274058.2 | NP_001260987.1 | 6 - 7 | 1591 | 71 | ||
NM_080153.5 | NP_524892.2 | 8 - 9 | 2074 | 71 | ||
GenBank mRNA | BT056247.1 | - | 8 - 9 | 2051 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |