Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273543.1 | NP_001260472.1 | 3 - 4 | 1498 | 139 | |
NM_080191.3 | NP_524930.1 | 3 - 4 | 1498 | 139 | ||
GenBank mRNA | AY051673.1 | - | 3 - 4 | 1466 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |