Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wun2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299319.1 | NP_001286248.1 | 5 - 6 | 1993 | 63 | |
NM_080252.4 | NP_524991.1 | 5 - 6 | 1750 | 63 | ||
GenBank mRNA | AF331162.1 | - | 5 - 6 | 1728 | 63 | |
BT050465.1 | - | 5 - 6 | 1748 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |