Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmet | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273451.1 | NP_001260380.1 | 8 - 9 | 5806 | 90 | |
NM_001273452.1 | NP_001260381.1 | 9 - 10 | 5962 | 90 | ||
NM_001273453.1 | NP_001260382.1 | 8 - 9 | 5806 | 90 | ||
NM_080254.3 | NP_524993.3 | 9 - 10 | 5971 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |