Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297877.1 | NP_001284806.1 | 4 - 5 | 2077 | 56 | |
NM_080304.3 | NP_525043.1 | 4 - 5 | 1192 | 56 | ||
GenBank mRNA | AY118763.1 | - | 4 - 5 | 1192 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |