Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272266.1 | NP_001259195.1 | 4 - 5 | 616 | 111 | |
NM_080318.3 | NP_525057.2 | 4 - 5 | 1200 | 111 | ||
GenBank mRNA | BT150201.1 | - | 4 - 5 | 1192 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |