Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop60B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014547.4 | NP_001014547.1 | 5 - 6 | 1391 | 69 | |
NM_001169815.3 | NP_001163286.1 | 5 - 6 | 1391 | 69 | ||
NM_001169818.3 | NP_001163289.1 | 5 - 6 | 1391 | 69 | ||
NM_080381.5 | NP_525120.1 | 5 - 6 | 1391 | 69 | ||
GenBank mRNA | DQ857345.1 | - | 5 - 6 | 1390 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |