Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fpps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058032.5 | NP_477380.1 | 5 - 6 | 1167 | 96 | |
GenBank mRNA | AF132554.1 | - | 5 - 6 | 1166 | 96 | |
AY071445.1 | - | 5 - 6 | 1162 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |