Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_164440.1 | NP_722733.1 | 1 - 2 | 1014 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Drosophila melanogaster |