Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GlyP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032048.2 | NP_001027219.1 | 3 - 4 | 2576 | 86 | |
NM_164453.4 | NP_722762.1 | 3 - 4 | 2576 | 86 | ||
GenBank mRNA | AF160947.1 | - | 3 - 4 | 2541 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |