Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Est-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274820.1 | NP_001261749.1 | 1 - 2 | 1431 | 98 | |
NM_176322.3 | NP_788500.1 | 1 - 2 | 1431 | 98 | ||
GenBank mRNA | BT099903.1 | - | 1 - 2 | 1427 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |