Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inaE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144723.2 | NP_001138195.1 | 9 - 10 | 2346 | 81 | |
NM_001144724.2 | NP_001138196.1 | 9 - 10 | 2346 | 81 | ||
NM_001298301.1 | NP_001285230.1 | 9 - 10 | 2346 | 81 | ||
NM_001298302.1 | NP_001285231.1 | 9 - 10 | 2346 | 81 | ||
NM_176727.3 | NP_788900.1 | 9 - 10 | 2259 | 81 | ||
NM_176728.3 | NP_788901.1 | 9 - 10 | 2346 | 81 | ||
GenBank mRNA | AY122194.1 | - | 9 - 10 | 2224 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |