Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201665.2 | NP_001188594.1 | 6 - 7 | 1164 | 92 | |
NM_001201666.2 | NP_001188595.1 | 6 - 7 | 1164 | 92 | ||
NM_001201667.2 | NP_001188596.1 | 6 - 7 | 1164 | 92 | ||
NM_001201668.2 | NP_001188597.1 | 5 - 6 | 1226 | 92 | ||
NM_001298379.1 | NP_001285308.1 | 6 - 7 | 1356 | 92 | ||
NM_176740.4 | NP_788913.2 | 6 - 7 | 1164 | 92 | ||
GenBank mRNA | AF222744.1 | - | 6 - 7 | 1146 | 92 | |
BT021400.1 | - | 6 - 7 | 1139 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |