Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275794.1 | NP_001262723.1 | 3 - 4 | 1141 | 89 | |
NM_057310.4 | NP_476658.1 | 2 - 3 | 1198 | 89 | ||
GenBank mRNA | BT132708.1 | - | 2 - 3 | 1174 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |