Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260338.2 | NP_001247267.1 | 7 - 8 | 3102 | 77 | |
NM_057796.4 | NP_477144.1 | 7 - 8 | 3102 | 77 | ||
GenBank mRNA | AF007805.1 | - | 7 - 8 | 3087 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |