Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Doa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014678.3 | NP_001014678.1 | 6 - 7 | 2418 | 103 | |
NM_001014679.2 | NP_001014679.1 | 4 - 5 | 1781 | 103 | ||
NM_001014680.4 | NP_001014680.1 | 7 - 8 | 1134 | 103 | ||
NM_001014681.4 | NP_001014681.2 | 6 - 7 | 5399 | 103 | ||
NM_001014682.3 | NP_001014682.1 | 7 - 8 | 1010 | 103 | ||
NM_001043298.2 | NP_001036763.2 | 5 - 6 | 3844 | 103 | ||
NM_001043299.3 | NP_001036764.2 | 8 - 9 | 1217 | 103 | ||
NM_001043300.3 | NP_001036765.1 | 7 - 8 | 1134 | 103 | ||
NM_001144648.3 | NP_001138120.1 | 8 - 9 | 1077 | 103 | ||
NM_001144649.2 | NP_001138121.1 | 8 - 9 | 1077 | 103 | ||
NM_001144650.1 | NP_001138122.1 | 7 - 8 | 1010 | 103 | ||
NM_001260424.1 | NP_001247353.1 | 5 - 6 | 2055 | 103 | ||
NM_001260425.1 | NP_001247354.1 | 6 - 7 | 2046 | 103 | ||
NM_001382085.1 | NP_001368951.1 | 8 - 9 | 1274 | 103 | ||
NM_176581.4 | NP_788758.3 | 6 - 7 | 2808 | 103 | ||
GenBank mRNA | AY061474.1 | - | 7 - 8 | 1128 | 103 | |
AY069573.1 | - | 4 - 5 | 1781 | 103 | ||
BT133441.1 | - | 6 - 7 | 2416 | 103 | ||
EF489455.1 | - | 6 - 7 | 2416 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |