Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057959.3 | NP_477307.1 | 3 - 4 | 273 | 72 | |
NM_169070.2 | NP_730952.1 | 3 - 4 | 273 | 72 | ||
NM_169072.3 | NP_730954.2 | 3 - 4 | 273 | 72 | ||
GenBank mRNA | AF077837.1 | - | 3 - 4 | 259 | 72 | |
AF160900.1 | - | 3 - 4 | 252 | 72 | ||
AF465756.1 | - | 2 - 3 | 140 | 72 | ||
GU452673.1 | - | 3 - 4 | 228 | 72 | ||
GU452674.1 | - | 3 - 4 | 225 | 72 | ||
GU452675.1 | - | 3 - 4 | 233 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |