Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058053.4 | NP_477401.2 | 5 - 6 | 2679 | 72 | |
NM_169430.3 | NP_731633.2 | 6 - 7 | 2619 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |