Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tacc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202238.2 | NP_001189167.1 | 3 - 4 | 415 | 80 | |
NM_001202239.2 | NP_001189168.1 | 5 - 6 | 3128 | 80 | ||
NM_001202240.2 | NP_001189169.1 | 5 - 6 | 3522 | 80 | ||
NM_001259986.1 | NP_001246915.1 | 5 - 6 | 3049 | 80 | ||
NM_001275332.1 | NP_001262261.1 | 5 - 6 | 3522 | 80 | ||
NM_001275333.1 | NP_001262262.1 | 6 - 7 | 3363 | 80 | ||
NM_058085.5 | NP_477433.2 | 5 - 6 | 3480 | 80 | ||
NM_058086.4 | NP_477434.1 | 5 - 6 | 3167 | 80 | ||
NM_169016.3 | NP_730867.2 | 5 - 6 | 3170 | 80 | ||
NM_176401.2 | NP_788578.1 | 5 - 6 | 3435 | 80 | ||
NM_176402.3 | NP_788579.1 | 3 - 4 | 373 | 80 | ||
NM_176403.2 | NP_788580.1 | 3 - 4 | 366 | 80 | ||
NM_176404.2 | NP_788581.1 | 3 - 4 | 408 | 80 | ||
GenBank mRNA | AY058499.1 | - | 5 - 6 | 3480 | 80 | |
BT099701.1 | - | 5 - 6 | 3136 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |