Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alpha-Est4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079542.4 | NP_524266.1 | 2 - 3 | 349 | 84 | |
GenBank mRNA | AY070981.1 | - | 2 - 3 | 324 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |