Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275482.1 | NP_001262411.1 | 13 - 14 | 2803 | 70 | |
NM_001275483.1 | NP_001262412.1 | 13 - 14 | 2806 | 70 | ||
NM_001382200.1 | NP_001368998.1 | 6 - 7 | 1294 | 70 | ||
NM_079564.4 | NP_524288.3 | 13 - 14 | 4165 | 70 | ||
NM_169274.2 | NP_731341.1 | 12 - 13 | 2602 | 70 | ||
NM_169275.2 | NP_731342.1 | 10 - 11 | 2043 | 70 | ||
NM_169276.2 | NP_731343.1 | 6 - 7 | 1510 | 70 | ||
GenBank mRNA | BT003462.1 | - | 13 - 14 | 4163 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |