Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alpha-Man-IIa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079567.3 | NP_524291.2 | 1 - 2 | 257 | 90 | |
GenBank mRNA | AY119464.1 | - | 1 - 2 | 190 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |