Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgf-D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079621.3 | NP_524345.2 | 2 - 3 | 633 | 76 | |
GenBank mRNA | AF337553.1 | - | 2 - 3 | 614 | 76 | |
BT010219.1 | - | 2 - 3 | 688 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |