Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cher | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202309.1 | NP_001189238.1 | 11 - 12 | 4380 | 66 | |
NM_001275728.1 | NP_001262657.2 | 13 - 14 | 5257 | 66 | ||
NM_001275729.1 | NP_001262658.1 | 12 - 13 | 4947 | 66 | ||
NM_001275730.1 | NP_001262659.2 | 14 - 15 | 5272 | 66 | ||
NM_079659.4 | NP_524383.3 | 11 - 12 | 4441 | 66 | ||
NM_206502.3 | NP_996224.2 | 12 - 13 | 5008 | 66 | ||
GenBank mRNA | AF188360.1 | - | 11 - 12 | 4417 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |