Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhc93AB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275864.1 | NP_001262793.1 | 7 - 8 | 2408 | 72 | |
NM_079700.3 | NP_524424.2 | 7 - 8 | 2408 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |