Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170197.1 | NP_001163668.1 | 9 - 10 | 3848 | 60 | |
NM_001260292.1 | NP_001247221.1 | 10 - 11 | 1732 | 60 | ||
NM_001275866.1 | NP_001262795.1 | 10 - 11 | 1719 | 60 | ||
NM_079701.5 | NP_524425.2 | 10 - 11 | 2035 | 60 | ||
NM_169944.3 | NP_732584.1 | 10 - 11 | 1719 | 60 | ||
NM_206529.2 | NP_996251.2 | 10 - 11 | 2035 | 60 | ||
GenBank mRNA | AY058543.1 | - | 10 - 11 | 1994 | 60 | |
BT044567.1 | - | 9 - 10 | 3846 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |