Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rtet | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079705.3 | NP_524429.1 | 5 - 6 | 1500 | 106 | |
GenBank mRNA | AF181656.1 | - | 5 - 6 | 1452 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |