Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABA-B-R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300527.1 | NP_001287456.1 | 2 - 3 | 362 | 75 | |
NM_079714.2 | NP_524438.1 | 2 - 3 | 362 | 75 | ||
NM_169968.1 | NP_732658.1 | 2 - 3 | 362 | 75 | ||
GenBank mRNA | AF145639.1 | - | 2 - 3 | 356 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |