Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300530.1 | NP_001287459.1 | 3 - 4 | 1204 | 72 | |
NM_079722.4 | NP_524446.3 | 3 - 4 | 1204 | 72 | ||
NM_169991.2 | NP_732694.2 | 4 - 5 | 1258 | 72 | ||
GenBank mRNA | AY071402.1 | - | 4 - 5 | 1258 | 72 | |
AY119620.1 | - | 3 - 4 | 1180 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |