Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTA1-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144539.2 | NP_001138011.1 | 8 - 9 | 2813 | 59 | |
NM_001275364.2 | NP_001262293.1 | 10 - 11 | 2468 | 59 | ||
NM_141309.4 | NP_649566.2 | 9 - 10 | 2360 | 59 | ||
NM_169086.3 | NP_730976.1 | 10 - 11 | 2468 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |