Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xe7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275367.1 | NP_001262296.1 | 5 - 6 | 2355 | 68 | |
NM_141315.4 | NP_649572.4 | 5 - 6 | 2272 | 68 | ||
NM_169093.3 | NP_730984.1 | 5 - 6 | 2272 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |