Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osi12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141374.1 | NP_649631.1 | 1 - 2 | 651 | 132 | |
GenBank mRNA | BT009951.1 | - | 1 - 2 | 650 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |